Skip to Content
Merck
All Photos(1)

Key Documents

MABS1254

Sigma-Aldrich

Anti-O-GlcNAc Antibody, clone CTD110.6

clone CTD110.6, from mouse

Synonym(s):

O-GlcNAc, beta-O-GlcNAc, O-Linked N-Acetylglucosamine, beta-O-linked N-acetylglucosamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$565.00

$565.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$565.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$565.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CTD110.6, monoclonal

species reactivity (predicted by homology)

all

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgMκ

target post-translational modification

glycosylation

Gene Information

human ... OGT(8473)

General description

Posttranslational modification of proteins by β-linked N-acetylglucosamine (β-GlcNAc) via the hydroxyl moieties on serine or threonine residues is termed O-linked β-GlcNAc or simply O-GlcNAc. O-GlcNAc is one of the most abundant posttranslational modifications within the nucleocytoplasmic compartments of all animals and plants. Unlike other types of protein glycosylations, O-GlcNAc occurs exclusively within the nuclear and cytoplasmic compartments and is generally not further modified to form more elongated structures. In addition, O-GlcNAcylation is a highly dynamic and reversible process. The O-GlcNAc transferase (OGT) attaches O-GlcNAc to proteins at specific serine or threonine residues, while O-GlcNAcase catalyzes the removal/hydrolysis of O-GlcNAc from proteins. In fact, a dynamic interplay between O-GlcNAcylation and serine/threonine phosphorylation plays an important role in regulating cellular signaling. Tau and RNA polymerase II (Pol II) are two well known proteins that undergo modification by O-GlcNAcylation. In Alzheimer’s diseased human brains, tau becomes extensively phosphorylated and less O-GlcNAcylated. Similarly, O-GlcNAc is removed and replaced with O-phosphate on the Poly II CTD when the elongation phase of transcription is initiated.

Specificity

Clone CTD110.6 specifically detected serine and threonine residues with beta-O-linked GlcNAc, but not alpha-O-linked GlcNAc. The presence of GlcNAc, but not GalNAc, abolished the detection of the target modification. Clone CTD110.6 did not recognize peptides with unmodified serine and threonine residues (Comer, F.I., et al. (2001). Anal. Biochem. 293(2):169-177).
Target modification is not species-specific.

Immunogen

Epitope: beta-O-linked GlcNAc.
KLH-conjugated peptide with an O-GlcNAc-modified serine residue.

Application

Anti-O-GlcNAc Antibody, clone CTD110.6 is an antibody against O-GlcNAc for use in Western Blotting, ELISA, Immunoprecipitation.
Research Category
Signaling
Research Sub Category
General Post-translation Modification
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected O-GlcNAcylated proteins in 7.5-15 µg of wild-type mouse embryonic fibroblast (MEF) lysate, but not O-GlcNAc transferase/OGT-deficient MEF lysate (Courtesy of Dr. Natasha Zachara and Gokben Yildirir, M.S.).
ELISA Analysis: A representative lot detected RNA polymerase II C-terminal domain (CTD) peptide (YSPTSPS) with a single O-GlcNAcylated serine or threonine, but not the corresponding unmodified peptide (Comer, F.I., et al. (2001). Anal. Biochem. 293(2):169-177).
Immunoprecipitation Analysis: A representative lot immunoprecipitated O-GlcNAcylated proteins from human pluripotent stem cells (hPSCs) (Maury, J.J., et al. (2013). Stem Cell Res. 11(2):926-937).
Immunoprecipitation Analysis: A representative lot immunoprecipitated O-GlcNAcylated proteins from HeLa cell extracts (Comer, F.I., et al. (2001). Anal. Biochem. 293(2):169-177).
Western Blotting Analysis: A representative lot detected similar level of cellular O-GlcNAcylation in undifferentiated, differentiating and terminally differentiated human pluripotent stem cells (hPSCs) (Maury, J.J., et al. (2013). Stem Cell Res. 11(2):926-937).
Western Blotting Analysis: A representative lot detected BSA-conjugated RNA polymerase II C-terminal domain (CTD) peptide (YSPTSPS) with beta-O-linked GlcNAc, but not alpha-O-linked GlcNAc, nor the corresponding unmodified peptide. The presence of GlcNAc, but not GalNAc, abolished the detection of the target bands (Comer, F.I., et al. (2001). Anal. Biochem. 293(2):169-177).
Western Blotting Analysis: A representative lot detected O-GlcNAcylated proteins in HeLa nuclear extract, as well as O-GlcNAcylated proteins purified from HeLa nuclear & cytosolic extract by wheat germ agglutinin (WGA) column. Antibody blocking by mmunogen peptide prior to immunoblotting abolished target bands detection (Comer, F.I., et al. (2001). Anal. Biochem. 293(2):169-177).
Western Blotting Analysis: A representative lot detected an upregulation of O-GlcNAcylated proteins in Jurkat cells treated with the glucosaminidase inhibitor PUGNAc and the hexosamine pathway intermediate glucosamine (Comer, F.I., et al. (2001). Anal. Biochem. 293(2):169-177).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 4.0 µg/mL of this antibody detected O-GlcNAcylated proteins in 10 µg of HeLa cell lysate.

Target description

Variable, depending on the size(s) of the O-GlcNAcylated protein(s).

Physical form

Format: Purified
Purified mouse monoclonal IgMκ antibody in PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giang Le Minh et al.
Frontiers in oncology, 13, 1141834-1141834 (2023-05-08)
Breast tumor development is regulated by a sub-population of breast cancer cells, termed cancer stem-like cells (CSC), which are capable of self-renewing and differentiating, and are involved in promoting breast cancer invasion, metastasis, drug resistance and relapse. CSCs are highly
Manuel González-Cuesta et al.
Journal of the American Chemical Society, 144(2), 832-844 (2022-01-06)
Owing to its roles in human health and disease, the modification of nuclear, cytoplasmic, and mitochondrial proteins with O-linked N-acetylglucosamine residues (O-GlcNAc) has emerged as a topic of great interest. Despite the presence of O-GlcNAc on hundreds of proteins within
Yaowen Zhang et al.
Journal of genetics and genomics = Yi chuan xue bao, 50(12), 948-959 (2023-06-08)
Protein O-GlcNAcylation is a monosaccharide post-translational modification maintained by two evolutionarily conserved enzymes, O-GlcNAc transferase (OGT) and O-GlcNAcase (OGA). Mutations in human OGT have recently been associated with neurodevelopmental disorders, although the mechanisms linking O-GlcNAc homeostasis to neurodevelopment are not
Meng-Jie Zhao et al.
EMBO reports, 22(6), e51649-e51649 (2021-04-16)
Pathological TDP-43 aggregation is characteristic of several neurodegenerative diseases, including amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration (FTLD-TDP); however, how TDP-43 aggregation and function are regulated remain poorly understood. Here, we show that O-GlcNAc transferase OGT-mediated O-GlcNAcylation of TDP-43
Kamau Fahie et al.
Current protocols, 1(5), e129-e129 (2021-05-19)
O-GlcNAc is a common post-translational modification of nuclear, mitochondrial, and cytoplasmic proteins that regulates normal physiology and the cell stress response. Dysregulation of O-GlcNAc cycling is implicated in the etiology of type II diabetes, heart failure, hypertension, and Alzheimer's disease

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service