Skip to Content
Merck
All Photos(3)

Key Documents

SAB5200053

Sigma-Aldrich

Monoclonal Anti-GABRA1 antibody produced in mouse

clone S96-55, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-AW061132, Anti-GABAARALPHA1 protein, S95-35, Anti-Gabrb-1, Anti-Gabrb1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$482.00

$482.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1626


Select a Size

Change View
100 μG
$482.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$482.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1626

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S96-55, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 55 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Gabra1(14394)

Specificity

Detects ~55 kDa. No cross-reactivity against GABA-A-R-Alpha 2 or –Alpha3.

Immunogen

Fusion protein amino acids 355-394 of mouse GABA-A-R-Alpha1, Accession Number P62812

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in PBS, pH 7.4, 50% glycerol, and 0.09% sodium azide

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service