Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700535

Sigma-Aldrich

Monoclonal Anti-CD263, azide free antibody produced in mouse

clone TRAIL-R3-02, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-TNFRSF10C, Anti-TRAIL-R3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$259.00

$259.00


Estimated to ship on04 May 2025



Select a Size

Change View
100 μG
$259.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$259.00


Estimated to ship on04 May 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TRAIL-R3-02, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

The antibody TRAIL-R3-02 reacts with TRAIL-R3, a 35 kDa GPI-anchored extracellular membrane protein expressed mainly on neutrophils.

Immunogen

TRAIL-R3 (aa 1-280) - hIgGhc fusion protein

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 1-5 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in azide free phosphate buffered saline, pH 7.4; 0.2 um filter sterilized.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service