Skip to Content
Merck
All Photos(2)

Key Documents

SAB4504372

Sigma-Aldrich

Anti-phospho-Cortactin (pTyr421) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$529.00

$529.00


Check Cart for Availability


Select a Size

Change View
100 μG
$529.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$529.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 61 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr421)

Gene Information

human ... CTTN(2017)

General description

Cortactin is an actin-related protein 2/3 (Arp2/3) complex–activating and filamentous (F)-actin–binding protein. This protein is coded by CTTN gene that maps to human chromosome 11q13.

Immunogen

The antiserum was produced against synthesized peptide derived from human Cortactin around the phosphorylation site of Tyr421.

Immunogen Range: 388-437

Application

Anti-phospho-Cortactin (pTyr421) antibody has been used in western blotting and immunoprecipitation.

Biochem/physiol Actions

Cortactin controls Arp2/3-mediated actin polymerization. This protein also modulates aggressive cancer cell traits like tumor growth, cancer cell invasion and motility, and serum- and anchorage-independent growth.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

β3 integrin expression is required for invadopodia-mediated ECM degradation in lung carcinoma cells.
Pelaez R, et al.
PLoS ONE, 12(8), e0181579-e0181579 (2017)
Cyclic expression of the voltage?gated potassium channel KV10. 1 promotes disassembly of the primary cilium.
Sanchez A, et al.
EMBO Reports, e201541082-e201541082 (2016)
Araceli Sánchez et al.
EMBO reports, 17(5), 708-723 (2016-04-27)
The primary cilium, critical for morphogenic and growth factor signaling, is assembled upon cell cycle exit, but the links between ciliogenesis and cell cycle progression are unclear. KV10.1 is a voltage-gated potassium channel frequently overexpressed in tumors. We have previously
Cortactin contributes to the tumorigenicity of colorectal cancer by promoting cell proliferation.
Wu H, et al.
Oncology Reports, 36(6), 3497-3503 (2016)
Cortactin promotes exosome secretion by controlling branched actin dynamics.
Sinha S, et al.
The Journal of Cell Biology, 214(2), 197-213 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service