Skip to Content
Merck
All Photos(1)

Key Documents

420314

Sigma-Aldrich

Kallikrein Substrate, Fluorogenic

≥95% (HPLC), lyophilized, kallikrein substrate, Calbiochem®

Synonym(s):

Kallikrein Substrate, Fluorogenic

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$311.00

$311.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$311.00

About This Item

Empirical Formula (Hill Notation):
C27H38F3N7O5 · 2HCl
Molecular Weight:
670.55
UNSPSC Code:
12352200
NACRES:
NA.77

$311.00


Estimated to ship on29 May 2025


Request a Bulk Order

Product Name

Kallikrein Substrate, Fluorogenic, Sensitive fluorogenic substrate for kallikreins.

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

solubility

5% acetic acid: 1 mg/mL
DMSO: 5 mg/mL

fluorescence

λex 400 nm
λem 505 nm

shipped in

ambient

storage temp.

−20°C

General description

Sensitive fluorogenic substrate for kallikreins (excitation maximum: 400 nm; emission maximum: 505 nm).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Fluorogenic substrate for kallikreins
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

H-D-Val-Leu-Arg-AFC, 2HCl

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Garrett, J.R., et al. 1985. Histochem. J.17, 805.
Takada, Y., et al. 1985. Biochem. J.232, 851.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service