Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501406

Sigma-Aldrich

Anti-Nuclear Receptor NR4A1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Early response protein NAK1, Nuclear receptor subfamily 4 group A member 1, Orphan nuclear receptor HMR, ST-59, TR3 orphan receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$693.00

$693.00


Check Cart for Availability


Select a Size

Change View
100 μG
$693.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$693.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 64 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR4A1(3164)

General description

Nuclear receptor subfamily 4 group A member1 (NR4A1) is an orphan receptor that belongs to the nuclear hormone receptor superfamily. The NR4A1 gene is mapped to human chromosome 12q13.13. Structurally, NR4A1 comprises an N-terminal AB region, DNA binding domain (DBD), and a C-terminal ligand-binding domain (LBD). Anti-Nuclear Receptor NR4A1 antibody detects endogenous levels of total Nuclear Receptor NR4A1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Nuclear Receptor NR4A1.

Immunogen Range: 317-366

Biochem/physiol Actions

Nuclear receptor subfamily 4 group A member1 (NR4A1) participates in gene expression regulation in the hypothalamic-pituitary-adrenal axis (HPA) especially those related to inflammation and steroidogenesis. The levels of NR4A1 are downregulated in metastatic melanoma, myelogenous leukemia, and follicular thyroid carcinoma. An elevated expression of NR4A1 is observed in type 1 diabetes mellitus (T1DM) and type 2 diabetes mellitus (T2DM) in mouse models. Dysfunction of NR4A1 is correlated to energy balance and also the events of metabolic pathways including carbohydrates and lipids. NR4A1 contributes to arterial vessel cell wall damage. It regulates tumor cell proliferation, migration, and invasion. NR4A1 may serve as a biomarker for depression and other psychiatric disorders.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transcripts of orphan nuclear receptor (NR4A1) & potassium channel (KCNK17) genes as new potential biomarkers for depression
Dmitrzak-Weglarz M, et al.
Meta Gene null
Stephen Safe et al.
The Journal of steroid biochemistry and molecular biology, 157, 48-60 (2015-04-29)
The orphan nuclear receptors NR4A1, NR4A2 and NR4A3 are immediate early genes induced by multiple stressors, and the NR4A receptors play an important role in maintaining cellular homeostasis and disease. There is increasing evidence for the role of these receptors
Lei Zhang et al.
Journal of diabetes research, 2018, 9363461-9363461 (2018-07-18)
Orphan nuclear receptor 4A1 (NR4A1) is a transcriptional factor of the nuclear orphan receptor (NR4A) superfamily that has sparked interest across different research fields in recent years. Several studies have demonstrated that ligand-independent NR4A1 is an immediate-early response gene and
K D Senali Abayratna Wansa et al.
The Journal of biological chemistry, 277(36), 33001-33011 (2002-06-26)
Nur77/NR4A1 is an "orphan member" of the nuclear hormone receptor superfamily. Nur77 and its close relatives Nurr1 and NOR-1 bind as monomers to a consensus binding site, the nerve growth factor induced protein I-B (NGFI-B)-binding response element (NBRE). The Nur77/NURR1/NOR1
Shaoxi Liao et al.
Genes, chromosomes & cancer, 51(12), 1067-1078 (2012-08-14)
Only a minority of intraductal carcinomas of the breast give rise to stromally invasive disease. We microdissected 206 paraffin blocks representing 116 different cases of low-grade ductal carcinoma in situ (DCIS). Fifty-five were pure DCIS (PD) cases without progression to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service