Skip to Content
Merck
All Photos(1)

Key Documents

MABF936

Sigma-Aldrich

Anti-Regnase-1/Zc3h12a Antibody, clone 15D11

clone 15D11, from rat

Synonym(s):

Ribonuclease ZC3H12A, MCP-induced protein 1, Regnase-1, Zinc finger CCCH domain-containing protein 12A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$563.00

$563.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$563.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$563.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

15D11, monoclonal

species reactivity

mouse

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Ribonuclease ZC3H12A (UniProt Q5D1E7; also known as MCP-induced protein 1, Regnase-1, Zinc finger CCCH domain-containing protein 12A) is encoded by the Zc3h12a (also known as Mcpip, Mcpip1) gene (Gene ID 230738) in murine species. Regnase-1 is an endonuclease that is involved in post-transcriptional gene regulation by cleaving target mRNAs or the terminal loop of pre-microRNAs. Regnase-1 negatively regulates Il6, Il12b, Il2, Icos, Ox40, Ctla4, cRel mRNAs, as well as its own mRNA (Zc3h12a). In T cells, regnase-1 and an RNA-binding protein roquin act together to inhibit T helper (TH) cell differentiation by negatively regulating mRNAs encoding the TH17 cell-promoting factors IL-6, ICOS, c-Rel, IRF4, IκBNS and IκBζ. This cooperation requires RNA-binding activity of roquin and the endonuclease activity of regnase-1. Upon T-cell receptor (TCR) activation following antigen binding, however, both roquin and regnase-1 are cleaved and inactivated by the paracaspase MALT1, allowing TH17 differentiation to proceed.

Specificity

Clone 15D11 detected regnase-1 in wild-type, but not Zc3h12a-/-, mouse embryonic fibroblasts/MEFs (Jeltsch, K.M., et al. (2014). Nat. Immunol. 15(11):1079-1089).

Immunogen

Recombinant full-length mouse regnase-1.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-Regnase-1/Zc3h12a Antibody, clone 15D11 is validated for use in Western Blotting for the detection of Regnase-1/Zc3h12a.
Western Blotting Analysis: A representative lot detected regnase-1 in lysates from wild-type, but not Zc3h12a-/-, mouse embryonic fibroblasts/MEFs (Jeltsch, K.M., et al. (2014). Nat. Immunol. 15(11):1079-1089).

Quality

Evaluated by Western Blotting in MEF1 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Regnase-1/Zc3h12a in 10 µg of MEF1 cell lysate.

Target description

~70 kDa observed.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service