Skip to Content
Merck
All Photos(3)

Key Documents

SAB4300188

Sigma-Aldrich

Anti-phospho-EIF4E (pSer209) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CBP antibody produced in rabbit, Anti-EIF4E1 antibody produced in rabbit, Anti-EIF4EL1 antibody produced in rabbit, Anti-EIF4F antibody produced in rabbit, Anti-eukaryotic translation initiation factor 4E antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$745.00

$745.00


Check Cart for Availability


Select a Size

Change View
100 μG
$745.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$745.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~25 kDa

species reactivity

mouse, human, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(S-G-SP-T-T)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer209)

Gene Information

human ... EIF4E(1977)

Immunogen

Peptide sequence around phosphorylation site of serine 209 (S-G-S(p)-T-T), according to the protein EIF4E.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Y Liu et al.
Journal of animal physiology and animal nutrition, 98(2), 310-317 (2013-05-31)
The objective of this study was to investigate the regulatory mechanism underlying the increased muscle protein accumulation in pigs while were fed a high protein diet. The eukaryotic initiation factors (eIFs) have been reported to involve in muscle protein synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service