Skip to Content
Merck
All Photos(1)

Key Documents

MAB1340

Sigma-Aldrich

Anti-Collagen Type I (COL1A1) Antibody

CHEMICON®, mouse monoclonal, C11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$521.00

$521.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$521.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$521.00


Check Cart for Availability

Request a Bulk Order

Product Name

Anti-Collagen Type I Antibody, clone C11, ascites fluid, clone C11, Chemicon®

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

C11, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... COL1A1(1277)

Specificity

Recognizes both native and heat denatured human collagen type I. No cross reactivity with albumin fractions of human serum or albumins of other animal species. Less than 5% binding to collagens II and II in ELISA, RIA or immunoblotting.

Application

Detect Collagen Type I using this Anti-Collagen Type I Antibody, clone C11 validated for use in ELISA, IH.
Immunohistochemical staining of 10 μm thick cryostat sections fixed with acetone or ethanol. Staining is similar to that for polyclonal antibodies to interstitial collagen I. Suggested working concentration 10-20 μg/mL. Formaldehyde fixation is not recommended. Does not work with Western blots. Not suitable for use on paraffin embedded sections.

ELISA: Apparent binding constant from titration curves is approximately 0.1nM. Antibody suitable for use in sandwich ELISA for specific determination of collagen I, employed as either a capture antibody or as a second, detecting antibody.

Optimal working dilutions must be determined by the end user.

Physical form

Immunoglobulin Purified from ascites by ammonium sulfate precipitation and chromatography on DEAE-cellulose. . Lyophilized, reconstitute with 1 mL of distilled water.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imaging cells and extracellular matrix in vivo by using second-harmonic generation and two-photon excited fluorescence.
Zoumi, A; Yeh, A; Tromberg, BJ
Proceedings of the National Academy of Sciences of the USA null
Patricia Khashayar et al.
3 Biotech, 7(5), 312-312 (2017-09-29)
This article explains a step-wise protocol to develop an electrochemical sensor to quantify serum levels of C-telopeptide (CTX) crosslinks also known as crosslaps in a matter of minutes and with high level of accuracy. The technique needs only one-step (incubation)
Jose M Gonzalez et al.
Scientific reports, 6, 21315-21315 (2016-02-18)
The contractile trabecular meshwork (TM) modulates aqueous humor outflow resistance and intraocular pressure. The primary goal was to visualize and quantify human TM contractile state by analyzing actin polymerization (F-actin) by 2-photon excitation fluorescence imaging (TPEF) in situ. A secondary
Human tendon stem cells better maintain their stemness in hypoxic culture conditions.
Zhang, J; Wang, JH
Testing null
Mesenchymal stem cells in rabbit meniscus and bone marrow exhibit a similar feature but a heterogeneous multi-differentiation potential: superiority of meniscus as a cell source for meniscus repair.
Ding, Z; Huang, H
BMC Musculoskelet. Disord. null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service