Skip to Content
Merck
All Photos(1)

Key Documents

SAB3700145

Sigma-Aldrich

Anti-Horse IgG (Fc specific)-Peroxidase antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

HRP

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
$460.00

$460.00


Check Cart for Availability


Select a Size

Change View
2 MG
$460.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$460.00


Check Cart for Availability

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

horse

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

The immunoglobulin IgG from horse comprises six subtypes. The subtypes share high sequence homology. Immunoglobulin super-family have a common structure, comprising two heavy (H) chains and two light (L) chains, held together by disulfide linkages. The heavy chain has one variable N-terminal region and three to four constant (CH1-CH4) C-terminal regions. The L chain comprises of one variable N-terminal region and a constant C-terminal region.

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Horse IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Peroxidase, Anti-Goat Serum, Horse IgG, Horse IgG F(c) and Horse Serum. No reaction was observed against Horse IgG F(ab′)2.

Immunogen

Horse IgG F(c) fragment

Biochem/physiol Actions

Immunoglobulin IgG digestion by papain results in the generation of fragment antigen binding (Fab). Pepsin digestion of IgG generates fragment crystallisable (Fc). The Fc region of IgG antibody has enormous therapeutic potential and is exploited for the development of therapeutic antibodies. Normal serum IgG levels (400-800 mg/dl) along with total serum proteins and serum globulin is essential in foals to alleviate the risk of developing failure of transfer of passive immunity(FTPI).

Physical properties

Antibody format: IgG

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2

Reconstitution

Reconstitute with 1.0 mL deionized water (or equivalent).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Usefulness of a commercial equine IgG test and serum protein concentration as indicators of failure of transfer of passive immunity in hospitalized foals
Metzger N, et al.
Journal of Veterinary Internal Medicine, 20(2), 382-387 (2006)
The complete map of the Ig heavy chain constant gene region reveals evidence for seven IgG isotypes and for IgD in the horse
Wagner B, et al.
Journal of Immunology, 173(5), 3230-3242 (2004)
Molecular properties of human IgG subclasses and their implications for designing therapeutic monoclonal antibodies against infectious diseases.
Irani V, et al.
Molecular Immunology, 67(2), 171-182 (2015)
Structure and function of immunoglobulins.
Schroeder Jr H W and Cavacini L
The Journal of Allergy and Clinical Immunology, 125(2), S41-S52 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service