Skip to Content
Merck
All Photos(1)

Documents

SAB3500444

Sigma-Aldrich

Anti-TIM-4 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-SMUCKLER, Anti-T cell immunoglobulin and mucin domain containing protein 4, Anti-T cell membrane protein 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TIMD4(91937)

Immunogen

TIM-4 antibody was raised against a 15 amino acid peptide from near the carboxy terminus of human TIM-4.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500444.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu Ding et al.
Immunology, 168(1), 49-62 (2022-08-01)
Obesity is generally associated with low-grade inflammation. Adipose tissue macrophages (ATMs) orchestrate metabolic inflammation. The classical (M1-like) or alternative (M2-like) activation of ATMs is functionally coupled with the metabolic status of fat tissues. It has been found that T-cell immunoglobulin-
Liyun Xu et al.
Laboratory investigation; a journal of technical methods and pathology, 96(11), 1189-1197 (2016-10-26)
Endotoxic shock is the primary cause of morbidity and mortality in hospital patients, creating an urgent need to explore the mechanisms involved in sepsis. Our previous studies showed that T-cell immunoglobulin- and mucin-domain-containing molecule-4 (Tim-4) attenuated the inflammatory response through
Wen Liu et al.
Journal of immunology (Baltimore, Md. : 1950), 203(4), 990-1000 (2019-07-03)
Nonalcoholic fatty liver disease (NAFLD), characterized by excessive inflammation and lipid deposition, is one of the most common metabolic liver diseases. The expression of NLRP3 inflammasome in macrophages is significantly increased in NAFLD, and its activation aggravates NAFLD greatly. Tim-4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service