Skip to Content
Merck
All Photos(2)

Key Documents

SAB3501241

Sigma-Aldrich

Anti-Integrin alpha 4 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$753.00

$753.00


Check Cart for Availability


Select a Size

Change View
100 μG
$753.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$753.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

human, mouse, rat

concentration

1 mg/mL

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITGA4(3676)

Looking for similar products? Visit Product Comparison Guide

General description

Integrin alpha 4 Antibody: The integrin alpha 4 (also known as CD49d and ITGA4) belongs to the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha and beta chains. Alpha4 chain associates with either beta1 or beta7 chain. It has been demonstrated that the putative ligand-binding sites of both integrin alpha4beta1 and alpha4beta7 is located on the alpha4 chain. These ligands included Madcam, VCAM, and fibronectin. Madcam is known as the principal ligand for integrin alpha4beta7. Recently it was also demonstrated that HIV-1 envelope can mimic Madcam by binding to and signaling though integrin alpha4beta7, the gut mucosal homing receptor for peripheral T cells.

Immunogen

Integrin alpha 4 antibody was raised against a 15 amino acid synthetic peptide from near the center of human Integrin alpha 4.The immunogen is located within amino acids 220 - 270 of Integrin alpha 4.

Application

Integrin alpha 4 antibody can be used for the detection of Integrin alpha 4 by Western blot at 1 μg/mL. Despite its predicted molecular weight, Integrin alpha4 will often run at higher molecular weight in SDS-PAGE. Antibody can also be used for immunoflourescence starting at 2 μg/mL. Antibody validated: Western Blot in rat samples and Immunofluorescence in human samples. All other applications and species not yet tested.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service