![Anti-Integrin alpha 4 antibody produced in rabbit affinity isolated antibody](/deepweb/assets/sigmaaldrich/product/images/147/623/d1a019af-1fbf-4aca-9131-ec15e496eceb/640/d1a019af-1fbf-4aca-9131-ec15e496eceb.jpg)
Anti-Integrin alpha 4 antibody produced in rabbit
![Anti-Integrin alpha 4 antibody produced in rabbit affinity isolated antibody](/deepweb/assets/sigmaaldrich/product/images/147/623/d1a019af-1fbf-4aca-9131-ec15e496eceb/640/d1a019af-1fbf-4aca-9131-ec15e496eceb.jpg)
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing