Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500896

Sigma-Aldrich

Anti-SARS SPIKE antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$470.00

$470.00


Estimated to ship on26 May 2025



Select a Size

Change View
100 μG
$470.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$470.00


Estimated to ship on26 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

yeast

concentration

1 mg/mL

technique(s)

ELISA: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Immunogen

Antibody was raised against a synthetic peptide corresponding to amino acids at the amino-terminus of. The sARS Spike glycoprotein.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500896.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenhui Li et al.
Nature, 426(6965), 450-454 (2003-12-04)
Spike (S) proteins of coronaviruses, including the coronavirus that causes severe acute respiratory syndrome (SARS), associate with cellular receptors to mediate infection of their target cells. Here we identify a metallopeptidase, angiotensin-converting enzyme 2 (ACE2), isolated from SARS coronavirus (SARS-CoV)-permissive
Marco A Marra et al.
Science (New York, N.Y.), 300(5624), 1399-1404 (2003-05-06)
We sequenced the 29,751-base genome of the severe acute respiratory syndrome (SARS)-associated coronavirus known as the Tor2 isolate. The genome sequence reveals that this coronavirus is only moderately related to other known coronaviruses, including two human coronaviruses, HCoV-OC43 and HCoV-229E.
Paul A Rota et al.
Science (New York, N.Y.), 300(5624), 1394-1399 (2003-05-06)
In March 2003, a novel coronavirus (SARS-CoV) was discovered in association with cases of severe acute respiratory syndrome (SARS). The sequence of the complete genome of SARS-CoV was determined, and the initial characterization of the viral genome is presented in
Sonia R Navas-Martín et al.
Journal of neurovirology, 10(2), 75-85 (2004-06-19)
A novel coronavirus has been recently identified as the causative agent of the severe acute respiratory syndrome (SARS) outbreak that has accounted for more than 8000 infected people worldwide. This review will discuss current knowledge on coronavirus replication, pathogenesis, evolution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service