Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500463

Sigma-Aldrich

Anti-gp120 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HIV-1 glycoprotein 120

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$721.00

$721.00


Check Cart for Availability


Select a Size

Change View
100 μG
$721.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$721.00


Check Cart for Availability

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

viral

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Immunogen

gp120 antibody was raised against 15 amino acids from near the center of gp120.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500463.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Wen Qian et al.
The Journal of biological chemistry, 291(44), 22977-22987 (2016-10-30)
Approximately 70% of HIV-1 infected patients acquire ocular opportunistic infections and manifest eye disorders during the course of their illness. The mechanisms by which pathogens invade the ocular site, however, are unclear. Under normal circumstances, vascular endothelium and retinal pigment
Ai-Ping Jiang et al.
Journal of virology, 90(6), 2928-2937 (2016-01-01)
The gastrointestinal mucosa is the primary site where human immunodeficiency virus type 1 (HIV-1) invades, amplifies, and becomes persistently established, and cell-to-cell transmission of HIV-1 plays a pivotal role in mucosal viral dissemination. Mast cells are widely distributed in the
Ai-Ping Jiang et al.
Journal of virology, 89(15), 8050-8062 (2015-05-29)
Cell-associated HIV-1 infection has been proposed to play a pivotal role in the spread of HIV-1 infection. Granulocytes are a category of white blood cells, comprising mainly basophils, neutrophils, and eosinophils, and participate in various inflammatory reactions and defense against

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service