Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500286

Sigma-Aldrich

Anti-LDL-R antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FH, Anti-FHC, Anti-LDLCQ2, Anti-LDLR, Anti-Low density lipoprotein receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$721.00

$721.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1176


Select a Size

Change View
100 μG
$721.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$721.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1176

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LDLR(3949)

Immunogen

LDL-R antibody was raised against an 18 amino acid peptide near the center of human LDL-R.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500286.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth A O'Hare et al.
Journal of lipid research, 55(11), 2242-2253 (2014-09-10)
Hyperlipidemia and arterial cholesterol accumulation are primary causes of cardiovascular events. Monogenic forms of hyperlipidemia and recent genome-wide association studies indicate that genetics plays an important role. Zebrafish are a useful model for studying the genetic susceptibility to hyperlipidemia owing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service