Skip to Content
Merck
All Photos(2)

Key Documents

SAB3500123

Sigma-Aldrich

Monoclonal Anti-PD-1 antibody produced in mouse

clone 12A7D7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CD279, Anti-PDCD-1, Anti-Programmed cell death 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$654.00

$654.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1025


Select a Size

Change View
100 μG
$654.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$654.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1025

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12A7D7, monoclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDCD1(5133)

General description

Programmed death-1 (PD-1) is a type I transmembrane protein, encoded by the gene mapped to human chromosome 2q37.3. PD-1 belongs to the immunoglobulin receptor superfamily. The encoded protein is an immunoinhibitory receptor expressed by activated T cells, B cells, and myeloid cells. PD-1 is characterized with a single Ig-like variable (IgV) domain in the extracellular region and an immunoreceptor tyrosine-based inhibitory domain in the cytoplasmic region.

Immunogen

raised against an ~150 amino acid recombinant protein from near the terminus terminus of mouse PD-1.

Biochem/physiol Actions

Programmed death-1 (PD-1) gene is sensitive to systemic lupus erythematosus (SLE). Polymorphisms in PD-1 may participate in the growth of EOC (epithelial ovarian cancer). It acts as a negative regulator of T cells. It is also responsible for immune escape mediated by tumor cells. Interaction of PD-1 and PD-L1 (ligand of PD-1) results in inhibition of T cell receptor–mediated lymphocyte proliferation and cytokine secretion.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of "danger signals" such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases. Despite its predicted molecular weight, PD-1 often migrates at higher molecular weight in SDS-PAGE.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service