Skip to Content
Merck
All Photos(3)

Key Documents

SAB2900809

Sigma-Aldrich

Anti-BCAS1 / NABC1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

AIBC1, BCAS1, NABC1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$763.00

$763.00


Check Cart for Availability


Select a Size

Change View
50 μG
$763.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$763.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

western blot: 1-2 μg/mL (Recombinant Protein)
western blot: 5 μg/mL (Cell Lysate)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BCAS1(8537)

Immunogen

BCAS1 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human BCAS1.
NABC1/BCAS1 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human BCAS1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan Yang et al.
Molecular brain, 12(1), 17-17 (2019-03-15)
Hypomyelination in the central nerves system (CNS) is one of the most obviously pathological features in Niemann-Pick Type C disease (NPC), which is a rare neurodegenerative disorder caused by mutations in the NPC intracellular cholesterol transporter 1 or 2 (Npc1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service