Skip to Content
Merck
All Photos(1)

Key Documents

MAB1578

Sigma-Aldrich

Anti-Muscarinic Acetylcholine Receptor m4 Antibody, clone 18C7.2

clone 18C7.2, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$711.00

$711.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$711.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$711.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

18C7.2, monoclonal

purified by

using Protein A

species reactivity

monkey

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

rhesus monkey ... Chrm4(574195)

Specificity

m4 muscarinic acetylcholine receptor. No reactivity with the other subtypes.

Immunogen

m4 receptor i3 loop (human), fused to GST

Application

Anti-Muscarinic Acetylcholine Receptor m4 Antibody, clone 18C7.2 detects level of Muscarinic Acetylcholine Receptor m4 & has been published & validated for use in IH, IP & WB.
Immunohistochemistry (monkey)

Western blot (transfected CHO cell lines)

Immunoprecipitation (rat and monkey)

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Physical form

Liquid in 0.02M Phosphate buffer, 0.25 M NaCl with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deborah Bauer et al.
Schizophrenia research, 117(1), 92-98 (2009-09-01)
The excitatory amino acid transporters (EAATs) are a family of molecules that are essential for regulation of synaptic glutamate levels. The EAATs may also be regulated by N-glycosylation, a posttranslational modification that is critical for many cellular functions including localization
G Q Li et al.
Neuroscience, 146(1), 384-402 (2007-03-30)
Muscarinic acetylcholine receptors (mAChRs) are widely expressed in the CNS and peripheral nervous system and play an important role in modulating the cell activity and function. We have shown that the cholinergic agonist carbachol reduces the pigeon's inwardly rectifying potassium
Christianne E Strang et al.
Investigative ophthalmology & visual science, 51(5), 2778-2789 (2010-01-01)
The activation and blockade of muscarinic acetylcholine receptors (mAChRs) affects retinal ganglion cell light responses and firing rates. This study was undertaken to identify the full complement of mAChRs expressed in the rabbit retina and to assess mAChR distribution and
Judith Molina et al.
Diabetes, 63(8), 2714-2726 (2014-03-25)
Acetylcholine regulates hormone secretion from the pancreatic islet and is thus crucial for glucose homeostasis. Little is known, however, about acetylcholine (cholinergic) signaling in the human islet. We recently reported that in the human islet, acetylcholine is primarily a paracrine
Jivan Khlghatyan et al.
Cerebral cortex (New York, N.Y. : 1991), 29(9), 3813-3827 (2018-10-09)
Cortical D2 dopamine receptor (Drd2) have mostly been examined in the context of cognitive function regulation and neurotransmission modulation of medial prefrontal cortex by principal neurons and parvalbumin positive, fast-spiking, interneurons in schizophrenia. Early studies suggested the presence of D2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service