Skip to Content
Merck
All Photos(3)

Key Documents

SAB2702069

Sigma-Aldrich

Anti-HHIP antibody produced in rabbit

Synonym(s):

HIP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$513.00

$513.00


Check Cart for Availability


Select a Size

Change View
100 μL
$513.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$513.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

79 kDa

species reactivity

chicken, human, mouse

concentration

0.86 mg/mL

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 500-3000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HHIP(64399)

General description

The gene encoding HHIP (hedgehog interacting protein) is located on human chromosome 4 and encodes for a protein belonging to the hedgehog-interacting protein (HHIP) family.[1]

Immunogen

Recombinant fragment corresponding to a region within amino acids 263 and 554 of HHIP (Uniprot ID#Q96QV1)

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Hedgehog interacting protein (HHIP) is highly conserved and interacts with all the three HH family members namely SHH (sonic hh), IHH (indian hh) and DHH (desert hh). It is also a vertebrate-specific inhibitor of HH signaling.[2] Single nucleotide polymorphisms in the HHIP gene are associated with increase in the risk of chronic obstructive pulmonary disease (COPD).[3]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of the Hedgehog signaling pathway in cancer: A comprehensive review.
Skoda AM, et al.
Bosnian Journal of Basic Medical Sciences / Udruzenje Basicnih Mediciniskih Znanosti = Association of Basic Medical Sciences, 18(1), 8-8 (2018)
Bo Wang et al.
Gene, 531(1), 101-105 (2013-09-03)
The hedgehog signaling pathway plays an important role in lung morphogenesis and cellular responses to lung injury. Genome-wide association studies (GWAS) and integrative genomics approaches have demonstrated the associations between HHIP polymorphisms and chronic obstructive pulmonary disease (COPD) and in
Xiaobo Zhou et al.
Genomics, 101(5), 263-272 (2013-03-06)
Hedgehog interacting protein (HHIP) was implicated in chronic obstructive pulmonary disease (COPD) by genome-wide association studies (GWAS). However, it remains unclear how HHIP contributes to COPD pathogenesis. To identify genes regulated by HHIP, we performed gene expression microarray analysis in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service