Skip to Content
Merck
All Photos(1)

Key Documents

SAB1407792

Sigma-Aldrich

Anti-PDP2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

KIAA1348

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$610.00

$610.00


Check Cart for Availability


Select a Size

Change View
50 μG
$610.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$610.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~60 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDP2(57546)

General description

Mouse polyclonal antibody raised against a full-length human PDP2 protein.
PDP2 (Pyruvate dehyrogenase phosphatase catalytic subunit 2) is a mitochondrial serine phos­phatase consisting of a catalytic subunit (PDP2c). It is a Mg2+ dependent enzyme. It has two active isoforms PDP 1 and 2. It is expressed in mammalian mitochondria.

Immunogen

PDP2 (NP_065837.1, 1 a.a. ~ 529 a.a) full-length human protein.

Sequence
MSSTVSYWILNSTRNSIATLQGGRRLYSRYVSNRNKLKWRLFSRVPPTLNSSPCGGFTLCKAYRHTSTEEDDFHLQLSPEQINEVLRAGETTHKILDLESRVPNSVLRFESNQLAANSPVEDRRGVASCLQTNGLMFGIFDGHGGHACAQAVSERLFYYVAVSLMSHQTLEHMEGAMESMKPLLPILHWLKHPGDSIYKDVTSVHLDHLRVYWQELLDLHMEMGLSIEEALMYSFQRLDSDISLEIQAPLEDEVTRNLSLQVAFSGATACMAHVDGIHLHVANAGDCRAILGVQEDNGMWSCLPLTRDHNAWNQAELSRLKREHPESEDRTIIMEDRLLGVLIPCRAFGDVQLKWSKELQRSILERGFNTEALNIYQFTPPHYYTPPYLTAEPEVTYHRLRPQDKFLVLASDGLWDMLSNEDVVRLVVGHLAEADWHKTDLAQRPANLGLMQSLLLQRKASGLHEADQNAATRLIRHAIGNNEYGEMEAERLAAMLTLPEDLARMYRDDITVTVVYFNSESIGAYYKGG

Application

Anti-PDP2 antibody produced in mouse is suitable for western blot analysis.

Biochem/physiol Actions

PDP2 (Pyruvate dehyrogenase phosphatase catalytic subunit 2) is indirectly associated with glycolysis and the tricarboxylic acid cycle and fatty acid (FA) synthesis. It is an Mg2+ dependent enzyme. PDP2 is mainly involved in the activation of phosphorylated pyruvate dehydrogenase complex (PDC), which is an essential component for the stimulation of the oxidative decarboxylation of pyruvate.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junko Kato et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 66(Pt 3), 342-345 (2010-03-09)
Pyruvate dehydrogenase phosphatase (PDP) is a mitochondrial serine phosphatase that activates phosphorylated pyruvate dehydrogenase complex by dephosphorylation. In humans, two PDP isoforms (1 and 2) have been identified. PDP1 is composed of a catalytic subunit (PDP1c) and a regulatory subunit
Mary C Sugden et al.
American journal of physiology. Endocrinology and metabolism, 284(5), E855-E862 (2003-04-05)
The mitochondrial pyruvate dehydrogenase complex (PDC) catalyzes the oxidative decarboxylation of pyruvate, linking glycolysis to the tricarboxylic acid cycle and fatty acid (FA) synthesis. Knowledge of the mechanisms that regulate PDC activity is important, because PDC inactivation is crucial for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service