Skip to Content
Merck
All Photos(4)

Key Documents

SAB2108342

Sigma-Aldrich

Anti-SGK1 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$716.00

$716.00


Estimated to ship on02 June 2025



Select a Size

Change View
100 μL
$716.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$716.00


Estimated to ship on02 June 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

49kDa

species reactivity

guinea pig, rabbit, bovine, human, mouse, rat, dog, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SGK1(6446)

General description

SGK1 (serum/glucocorticoid regulated kinase 1) codes for a serine/threonine kinase and it is structurally and functionally homologous to kinases of AKT (protein kinase B) family. SGK1 is ubiquitously expressed in all tissues and is predominant in kidney. The SKG1 gene is mapped to human chromosome 6q23.2.

Immunogen

Synthetic peptide directed towards the N terminal region of human SGK1

Biochem/physiol Actions

The SGK1 (serum/glucocorticoid regulated kinase 1) gene is associated with a number of pathophysiological processes such as autophagy, ion transport, proliferation, metabolic, inflammation, syndrome and endoplasmic reticulum stress. Cellular dehydration, increased extracellular salt concentration, hormones such as glucocorticoids, mineralocorticoids, transforming growth factor β and mediators (cytokines) such as interleukin -6, stimulates SGK1 action. SGK1 gene expression is regulated by cellular signals involving cytosolic Ca2+, cyclic AMP (adenosine monophosphate), stress-activated protein kinase (SAPK2 (serine/threonine-protein kinase 2), p38 kinase), protein kinase C. Also, insulin and insulin-like growth factor, ERK (extracellular signal-regulated kinase) 1/2, and hepatic growth factor induces SGK1 transcription. Upregulation of SGK1 is observed in diabetes, liver cirrhosis, glomerulonephritis, dialysis and several tumors.

Sequence

Synthetic peptide located within the following region: ANPSPPPSPSQQINLGPSSNPHAKPSDFHFLKVIGKGSFGKVLLARHKAE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Associations of the Serum/Glucocorticoid Regulated Kinase Genes With BP Changes and Hypertension Incidence: The Gensalt Study.
Zhang D, et al.
American Journal of Hypertension, 30(1), 95-101 (2016)
Lnc-SGK1 induced by Helicobacter pylori infection and highsalt diet promote Th2 and Th17 differentiation in human gastric cancer by SGK1/Jun B signaling.
Yao Y, et al.
Oncotarget, 7(15), 20549-20549 (2016)
SGK1 inhibits PM2. 5-induced apoptosis and oxidative stress in human lung alveolar epithelial A549 cells.
Li J, et al.
Biochemical and Biophysical Research Communications, 496(4), 1291-1295 (2018)
Single nucleotide polymorphism microarray analysis in cortisol-secreting adrenocortical adenomas identifies new candidate genes and pathways.
Ronchi C L, et al.
Neoplasia, 14(3), 206-218 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service