L1418
Anti-LAMP1 antibody produced in rabbit
affinity isolated antibody, buffered aqueous solution
Synonym(s):
LAMP1 Antibody - Anti-LAMP1 antibody produced in rabbit, Lamp1 Antibody, Anti-CD107a, Anti-LAMPA, Anti-LGP120, Anti-Lysosomal-associated membrane protein 1
Select a Size
About This Item
Recommended Products
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen ~120 kDa
species reactivity
rat, mouse, human
packaging
antibody small pack of 25 μL
technique(s)
indirect immunofluorescence: 5-10 μg/mL using human HeLa, rat NRK, and mouse NIH3T3 cells
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... LAMP1(3916)
mouse ... Lamp1(16783)
rat ... Lamp1(25328)
General description
Immunogen
Application
Biochem/physiol Actions
Physical form
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service