Skip to Content
Merck
All Photos(2)

Key Documents

SAB2106436

Sigma-Aldrich

Anti-EZH2 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$528.00

$528.00


Check Cart for Availability


Select a Size

Change View
100 μL
$528.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$528.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

85 kDa

species reactivity

guinea pig, horse, dog, rat, bovine, rabbit, human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... EZH2(2146)

Immunogen

Synthetic peptide directed towards the N terminal region of human EZH2

Biochem/physiol Actions

EZH2 is a member of the Polycomb-group (PcG) family. PcG family members form multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. This protein associates with the embryonic ectoderm development protein, the VAV1 oncoprotein, and the X-linked nuclear protein. This protein may play a role in the hematopoietic and central nervous systems.

Sequence

Synthetic peptide located within the following region: KGPVCWRKRVKSEYMRLRQLKRFRRADEVKSMFSSNRQKILERTEILNQE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ziming Li et al.
FEBS letters, 588(17), 3000-3007 (2014-06-15)
EZH2 is a key component of the polycomb PRC2 complex and functions as a histone H3 Lys27 (H3K27) trimethyltransferase. Here we show that EZH2 is down-regulated in human non-small cell lung cancer and low EZH2 expression predicts poor survival. Further
Shu-Qin Chen et al.
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society, 24(9), 1653-1658 (2014-10-03)
The current study was undertaken to investigate the predictive value of simultaneous enhancer of zeste homolog 2 (EZH2) and P53 expression in lesions of patients with cervical squamous cell carcinoma. Quantum dot double fluorescence staining was applied to detect EZH2
Jurate Savickiene et al.
Anti-cancer drugs, 25(8), 938-949 (2014-05-08)
Therapeutic strategies targeting histone deacetylase (HDAC) inhibition have become promising in many human malignancies. Belinostat (PXD101) is a hydroxamate-type HDAC inhibitor tested in phase I and II clinical trials in solid tumors and hematological cancers. However, little is known about

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service