Merck
  • The polycomb group protein EZH2 inhibits lung cancer cell growth by repressing the transcription factor Nrf2.
The polycomb group protein EZH2 inhibits lung cancer cell growth by repressing the transcription factor Nrf2.
FEBS letters (2014-06-15)
Ziming Li, Ling Xu, Naiwang Tang, Yunhua Xu, Xiangyun Ye, Shengping Shen, Xiaomin Niu, Shun Lu, Zhiwei Chen
ABSTRACT
EZH2 is a key component of the polycomb PRC2 complex and functions as a histone H3 Lys27 (H3K27) trimethyltransferase. Here we show that EZH2 is down-regulated in human non-small cell lung cancer and low EZH2 expression predicts poor survival. Further we demonstrate that EZH2 inhibits lung cancer cell proliferation and colony formation in vitro and growth in vivo. We found that EZH2 binds to the promoter of Nrf2, where it increases H3K27me3 and represses Nrf2 expression. Finally, Nrf2 seems to be essential for the hyper proliferation of lung cancer cells in the absence of EZH2.
Sigma-Aldrich
Anti-NFE2L2 antibody produced in rabbit
Sigma-Aldrich
Anti-EZH2 antibody produced in rabbit, affinity isolated antibody