Skip to Content
Merck
All Photos(4)

Key Documents

SAB2101144

Sigma-Aldrich

Anti-IKZF1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Hs.54452, Anti-IK1, Anti-IKAROS, Anti-IKAROS family zinc finger 1 (Ikaros), Anti-LYF1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$609.00

$609.00


Check Cart for Availability


Select a Size

Change View
100 μL
$609.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$609.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

57 kDa

species reactivity

human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IKZF1(10320)

General description

The Ikaros family zinc finger 1 (IKZF1) gene encodes for the Ikaros transcription factor, and is expressed throughout hematopoiesis. IKZF1 protein is made of an N-terminal DNA-binding domain (DBD) and a C-terminal dimerization domain. The IKZF1 gene is mapped on the human chromosome at 7p12.2.

Immunogen

Synthetic peptide directed towards the middle region of human IKZF1

Biochem/physiol Actions

Ikaros positively regulates lymphocyte differentiation and negatively regulates cell proliferation. It plays a role in peripheral lymphocyte homeostasis and negatively regulates follicular B cell activation. Ikaros/IKZF1 is involved in the development of myeloid cells such as red blood cells, platelets, basophils, and neutrophils. Mutations in the IKZF1 gene lead to acute lymphoblastic leukemia (ALL). Downregulation of the IKZF1 gene is observed in the peripheral blood mononuclear cells of systemic lupus erythematosus (SLE) patients. IKZF1 binds and activates the enhancer (delta-A element) of the CD3-delta gene. It functions in the specification and the maturation of the T-lymphocyte. It also interacts with a critical control element in the terminal deoxynucleotidyltransferase (TDT) promoter as well as with the promoters for other genes expressed during early stages of B- and T-cell development.

Sequence

Synthetic peptide located within the following region: DLCKIGSERSLVLDRLASNVAKRKSSMPQKFLGDKGLSDTPYDSSASYEK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles G Mullighan et al.
The New England journal of medicine, 360(5), 470-480 (2009-01-09)
Despite best current therapy, up to 20% of pediatric patients with acute lymphoblastic leukemia (ALL) have a relapse. Recent genomewide analyses have identified a high frequency of DNA copy-number abnormalities in ALL, but the prognostic implications of these abnormalities have
Su-juan Hu et al.
Modern rheumatology, 23(2), 205-209 (2012-07-12)
Ikaros family zinc finger 1, encoded by IKZF1, are lymphoid-restricted zinc finger transcription factors that share common N-terminal Kruppel-like zinc finger DNA-binding domain. IKZF1 play multiple important roles on regulators of lymphocyte differentiation and hematological tumor suppressor. Our genome-wide association
Agata Pastorczak et al.
Leukemia research, 35(11), 1534-1536 (2011-09-06)
Recent studies have shown that SNPs mapping to 7p12.2 (IKZF1), 9p21 (CDKN2A), 10q21.2 (ARID5B), and 14q11.2 (CEBPE) and carrier status for recessively inherited Nijmegen Breakage syndrome (NBS) influence childhood acute lymphoblastic leukemia (ALL) risk. To examine these relationship, we analysed
Biclonal and biallelic deletions occur in 20% of B-ALL cases with IKZF1 mutations.
A Dupuis et al.
Leukemia, 27(2), 503-507 (2012-08-08)
David Boutboul et al.
The Journal of clinical investigation, 128(7), 3071-3087 (2018-06-12)
Ikaros/IKZF1 is an essential transcription factor expressed throughout hematopoiesis. IKZF1 is implicated in lymphocyte and myeloid differentiation and negative regulation of cell proliferation. In humans, somatic mutations in IKZF1 have been linked to the development of B cell acute lymphoblastic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service