Skip to Content
Merck
All Photos(1)

Key Documents

MABE913

Sigma-Aldrich

Anti-Ikaros Antibody, clone Ik14

clone Ik14, from mouse

Synonym(s):

DNA-binding protein Ikaros, Ikaros family zinc finger protein 1, Lymphoid transcription factor LyF-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$657.00

$657.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$657.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$657.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Ik14, monoclonal

species reactivity

human

technique(s)

ChIP: suitable (ChIP-seq)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IKZF1(10320)

General description

Ikaros, also known as IKZF1, is a transcription factor that is important for the regulation of lymphopoiesis and also the development of the immune system. It has also been linked to adult and fetal erythroid differentiation in humans. Ikaros has been shown to induce local chromatin accessibility during CD4+ T cell differentiation and therefore, activating Th2 expression.

Immunogen

Recombinant protein corresponding to Human Ikaros.

Application

Anti-Ikaros, clone Ik14 is an antibody against Ikaros for use in WB, ChIP, ChIP-seq.
Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Chromatin Biology
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Ikaros in 10 µg of HL-60 and human lymph node lysate.
Chromatin Immunoprecipitation (ChIP): A representative lot of this antibody as part of a blend Ikaros clones Ik14 and E9 (catalogue numbers MABE913 and MABE912 respectively) was demonstrated to perform in ChIP. See Zhang et al. (2012).
ChIP-seq Analsysis: A representative lot of this antibody as part of a blend Ikaros clones E9 and Ik14 (catalogue number MABE912 and MABE913 respectively) was demonstrated to perform in ChIP-seq. See Zhang et al. (2012).

Quality

Evaluated by Western Blotting in a Jurkat cell lysate.

Western Blotting Analysis: A 0.5 µg/mL dilution of this antibody detected Ikaros in 10 µg of Jurkat cell lysate.

Target description

~60 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service