Skip to Content
Merck
All Photos(2)

Key Documents

SAB2101136

Sigma-Aldrich

Anti-IGF1R antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CD221, Anti-IGFIR, Anti-Insulin-like growth factor 1 receptor, Anti-JTK13, Anti-MGC142170

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$555.00

$555.00


Check Cart for Availability


Select a Size

Change View
100 μL
$555.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$555.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

71 kDa

species reactivity

pig, horse, sheep, bovine, dog, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IGF1R(3480)

General description

Insulin-like growth factor 1 receptor (IGF1R), a transmembrane tyrosine kinase receptor, belongs to the insulin receptor family. It comprises juxtamembrane (JM) and transmembrane domains that harbor substrate binding sites. Structurally, IGF1R exists as a tetramer α2/β2 with the extracellular dimeric α subunits. The IGF1R gene is mapped to human chromosome 15q26.3.

Immunogen

Synthetic peptide directed towards the middle region of human IGF1R

Biochem/physiol Actions

Insulin-like growth factor 1 receptor (IGF1R) is prime for IGF-1 for its mitogenic and metabolic functionality. It binds to IGF1 and IGF2 and activates phosphatidylinositol 3?kinase (PI3K)/protein kinase B (AKT) and mitogen-activated protein kinase pathways. IGF1R plays a key role in growth, differentiation, cell metabolic events, and apoptosis. It also favors proliferation in the myelodysplastic syndrome (MDS) and may serve as a potential target to treat MDS. Mutations in the IGF1R gene are implicated in the pre- and postnatal growth retardation and microcephaly. High levels of IGF1R overexpression are observed in multiple myeloma, lung, breast, bladder and pancreatic tumors.

Sequence

Synthetic peptide located within the following region: DRHSGHKAENGPGPGVLVLRASFDERQPYAHMNGGRKNERALPLPQSSTC

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi He et al.
Oncology reports, 44(3), 1094-1104 (2020-06-26)
Type 1 insulin‑like growth factor receptor (IGF‑IR) signaling is considered to serve a key role in the development of cancer. However, the effects of IGF‑IR on the malignant characteristics of myelodysplastic syndrome (MDS) clonal cells remains to be determined. In
Andrey S Kuznetsov et al.
Biochimica et biophysica acta. Biomembranes, 1862(11), 183417-183417 (2020-07-28)
Despite the biological significance of insulin signaling, the molecular mechanisms of activation of the insulin receptor (IR) and other proteins from its family remain elusive. Current hypothesis on signal transduction suggests ligand-triggered structural changes in the extracellular domain followed by
Nilda Gonzalez-Roibon et al.
Urology, 83(6), 1444-1444 (2014-04-10)
To assess the insulin-like growth factor-1 receptor (IGF1R) expression in urothelial carcinoma (UC) and its prognostic role in relation to clinicopathologic parameters. A total of 100 cases of invasive UC were evaluated using tissue microarrays. Membranous IGF1R staining was evaluated
Matias Juanes et al.
Clinical endocrinology, 82(5), 704-711 (2014-07-22)
IGF1R gene mutations have been associated with varying degrees of intrauterine and postnatal growth retardation, and microcephaly. To identify and characterize IGF1R gene variations in a cohort of 28 Argentinean children suspected of having IGF-1 insensitivity, who were selected on
Dan Tian et al.
BMC systems biology, 8, 98-98 (2014-08-15)
The insulin-like growth factor (IGF) system impacts cell proliferation and is highly activated in ovarian cancer. While an attractive therapeutic target, the IGF system is complex with two receptors (IGF1R, IGF2R), two ligands (IGF1, IGF2), and at least six high

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service