Skip to Content
Merck
All Photos(2)

Key Documents

SAB1412566

Sigma-Aldrich

ANTI-HNF4A antibody produced in mouse

clone 1F2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ39654, HNF4, HNF4A, HNF4a7, HNF4a8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$546.00

$546.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1457


Select a Size

Change View
100 μG
$546.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$546.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1457

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F2, monoclonal

form

buffered aqueous solution

mol wt

antigen 38.28 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HNF4A(3172)

General description

The protein encoded by this gene is a nuclear transcription factor which binds DNA as a homodimer. The encoded protein controls the expression of several genes, including hepatocyte nuclear factor 1 alpha, a transcription factor which regulates the expression of several hepatic genes. This gene may play a role in development of the liver, kidney, and intestines. Mutations in this gene have been associated with monogenic autosomal dominant non-insulin-dependent diabetes mellitus type I. Alternative splicing of this gene results in multiple transcript variants. (provided by RefSeq)

Immunogen

HNF4A (NP_849180, 351 a.a. ~ 464 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MIEQIQFIKLFGMAKIDNLLQEMLLGGSPSDAPHAHHPLHPHLMQEHMGTNVIVANTMPTHLSNGQMSTPETPQPSPPGGSGSEPYKLLPGAVATIVKPLSAIPQPTITKQEVI*

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service