Skip to Content
Merck
All Photos(1)

Key Documents

05-753

Sigma-Aldrich

Anti-WT1 Antibody, clone 6F-H2

clone 6F-H2, Upstate®, from mouse

Synonym(s):

Anti-AWT1, Anti-GUD, Anti-NPHS4, Anti-WAGR, Anti-WIT-2, Anti-WT-1, Anti-WT33

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
$660.00

$660.00


Estimated to ship on18 April 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1677

Request a Bulk Order

Select a Size

Change View
200 μG
$660.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$660.00


Estimated to ship on18 April 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1677

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6F-H2, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WT1(7490)

Specificity

WT1

Immunogen

6His-tagged fusion protein corresponding to residues 1-181 of human WT1 (Wilms tumor)

Application

Anti-WT1 Antibody, clone 6F-H2 is a Mouse Monoclonal Antibody for detection of WT1 also known as Wilms′ Tumor & has been tested in WB, ICC & IHC.

Quality

routinely evaluated by immunoblot on RIPA lysates from Jurkat and Raji cells

Target description

55-60kDa

Physical form

Format: Purified

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen Wu et al.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer, 8(9), 1163-1169 (2013-08-16)
The Wilms' tumor gene (WT1) has been identified as an oncogene in many malignant diseases, and aberrant WT1 expression has been linked to development, progression, and prognosis of non-small-cell lung cancer (NSCLC). We sought to investigate the underlying mechanism of
Rebecca Vicente-Steijn et al.
PloS one, 10(9), e0136025-e0136025 (2015-09-22)
Morphological and functional differences of the right and left ventricle are apparent in the adult human heart. A differential contribution of cardiac fibroblasts and smooth muscle cells (populations of epicardium-derived cells) to each ventricle may account for part of the
Zenglai Tan et al.
Scientific reports, 8(1), 16618-16618 (2018-11-11)
The kidney is a complex organ that is comprised of thousands of nephrons developing through reciprocal inductive interactions between metanephric mesenchyme (MM) and ureteric bud (UB). The MM undergoes mesenchymal to epithelial transition (MET) in response to the signaling from
Toru Sakairi et al.
American journal of physiology. Renal physiology, 298(3), F557-F567 (2009-12-04)
Evidence suggests that loss of podocytes into urine contributes to development of glomerular diseases; shed podocytes are frequently viable and proliferate in culture conditions. To determine the phenotypic characteristics of viable urinary cells derived from human subjects, we established long-term
Jing Zhou et al.
Biology open, 7(5) (2018-04-19)
Mesodermal populations can be generated in vitro from mouse embryonic stem cells (mESCs) using three-dimensional (3-D) aggregates called embryoid bodies or two-dimensional (2-D) monolayer culture systems. Here, we investigated whether Brachyury-expressing mesodermal cells generated using 3-D or 2-D culture systems

Questions

  1. On what buffer is the antibody dissolved?

    1 answer
    1. 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service