Skip to Content
Merck
All Photos(5)

Key Documents

SAB1412362

Sigma-Aldrich

ANTI-TBX18 antibody produced in mouse

clone 4D3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

TBX18

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$642.00

$642.00


Estimated to ship on29 May 2025



Select a Size

Change View
100 μG
$642.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$642.00


Estimated to ship on29 May 2025


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4D3, monoclonal

form

buffered aqueous solution

mol wt

antigen 37.4 kDa

species reactivity

, human,

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TBX18(9096)

General description

T-box transcription factor 18 (TBX18) belongs to the TBX family of transcription factors. It contains a conserved T-box domain and an N-terminal nuclear localization signal (NLS). The TBX18 gene is mapped to human chromosome 6q14.3

Immunogen

TBX18 (XP_496819, 454 a.a. ~ 560 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
STLLQGTGNGVPATHPHLLSGSSCSSPAFHLGPNTSQLCSLAPADYSACARSGLTLNRYSTSLAETYNRLTNQAGETFAPPRTPSYVGVSSSTSVNMSMGGTDGDTF

Application

ANTI-TBX18 antibody produced in mouse has been used in immunofluorescence staining.

Biochem/physiol Actions

T-box transcription factor 18 (TBX18) is involved in the sinoatrial node (SAN) formation and is expressed in the development and differentiation stages. It is a crucial factor for the pacemaker cell formation from cardiomyocytes. TBX18 may be implicated in congenital heart defects and congenital anomalies of the kidneys and urinary tract (CAKUT).

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergey Yechikov et al.
Stem cell research, 49, 102043-102043 (2020-11-01)
Directed cardiomyogenesis from human induced pluripotent stem cells (hiPSCs) has been greatly improved in the last decade but directed differentiation to pacemaking cardiomyocytes (CMs) remains incompletely understood. In this study, we demonstrated that inhibition of NODAL signaling by a specific
Henner F Farin et al.
The Journal of biological chemistry, 282(35), 25748-25759 (2007-06-23)
Tbox18 (Tbx18) and Tbox15 (Tbx15) encode a closely related pair of vertebrate-specific T-box (Tbx) transcription factors. Functional analyses in the mouse have proven the requirement of Tbx15 in skin and skeletal development and of Tbx18 in the formation of the
Aafke Engwerda et al.
European journal of human genetics : EJHG, 26(10), 1478-1489 (2018-06-16)
Proximal 6q (6q11-q15) deletions are extremely rare and little is known about their phenotypic consequences. Since parents and caregivers now use social media to seek information on rare disorders, the Chromosome 6 Project has successfully collaborated with a Facebook group
Soumya Negi et al.
Developmental biology, 446(2), 180-192 (2018-12-31)
The evolutionarily conserved transcription factor, Tbx18, is expressed in a dynamic pattern throughout embryonic and early postnatal life and plays crucial roles in the development of multiple organ systems. Previous studies have indicated that this dynamic function is controlled by
NODAL inhibition promotes differentiation of pacemaker-like cardiomyocytes from human induced pluripotent stem cells
Yechikov S, et al.
Stem Cell Research (2021)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service