Skip to Content
Merck
All Photos(1)

Key Documents

SAB1410594

Sigma-Aldrich

Anti-GTF2H2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

BTF2, BTF2P44, MGC102806, T-BTF2P44, TFIIH

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$583.00

$583.00


Check Cart for Availability


Select a Size

Change View
100 μG
$583.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$583.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 44.4 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GTF2H2(2966)

General description

This gene is part of a 500 kb inverted duplication on chromosome 5q13. This duplicated region contains at least four genes and repetitive elements which make it prone to rearrangements and deletions. The repetitiveness and complexity of the sequence have also caused difficulty in determining the organization of this genomic region. This gene is within the telomeric copy of the duplication. Deletion of this gene sometimes accompanies deletion of the neighboring SMN1 gene in spinal muscular atrophy (SMA) patients but it is unclear if deletion of this gene contributes to the SMA phenotype. This gene encodes the 44 kDa subunit of RNA polymerase II transcription initiation factor IIH which is involved in basal transcription and nucleotide excision repair. Transcript variants for this gene have been described, but their full length nature has not been determined. A second copy of this gene within the centromeric copy of the duplication has been described in the literature. It is reported to be different by either two or four base pairs; however, no sequence data is currently available for the centromeric copy of the gene. (provided by RefSeq)

Immunogen

GTF2H2 (NP_001506.1, 1 a.a. ~ 395 a.a) full-length human protein.

Sequence
MDEEPERTKRWEGGYERTWEILKEDESGSLKATIEDILFKAKRKRVFEHHGQVRLGMMRHLYVVVDGSRTMEDQDLKPNRLTCTLKLLEYFVEEYFDQNPISQIGIIVTKSKRAEKLTELSGNPRKHITSLKKAVDMTCHGEPSLYNSLSIAMQTLKHMPGHTSREVLIIFSSLTTCDPSNIYDLIKTLKAAKIRVSVIGLSAEVRVCTVLARETGGTYHVILDESHYKELLTHHVSPPPASSSSECSLIRMGFPQHTIASLSDQDAKPSFSMAHLDGNTEPGLTLGGYFCPQCRAKYCELPVECKICGLTLVSAPHLARSYHHLFPLDAFQEIPLEEYNGERFCYGCQGELKDQHVYVCAVCQNVFCVDCDVFVHDSLHCCPGCIHKIPAPSGV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service