Skip to Content
Merck
All Photos(3)

Key Documents

SAB1406349

Sigma-Aldrich

Anti-PTPRN2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

IA-2beta, IAR, ICAAR, PTPRP

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$691.00

$691.00


Check Cart for Availability


Select a Size

Change View
50 μG
$691.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$691.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~108.1 kDa

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTPRN2(5799)

General description

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domain, and thus represents a receptor-type PTP. The catalytic domain of this PTP is most closely related to PTPRN/IA-2beta. This PTP and PTPRN are both found to be major autoantigens associated with insulin-dependent diabetes mellitus. Three alternatively spliced transcript variants of this gene, which encode distinct proteins, have been reported. (provided by RefSeq)

Immunogen

PTPRN2 (NP_570858.1, 1 a.a. ~ 986 a.a) full-length human protein.

Sequence
MGPPLPLLLLLLLLLPPRVLPAAPSSVPRGRQLPGRLGCLLEEGLCGASEACVNDGVFGRCQKVPAMDFYRYEVSPVALQRLRVALQKLSGTGFTWQDDYTQYVMDQELADLPKTYLRRPEASSPARPSKHSVGSERRYSREGGAALANALRRHLPFLEALSQAPASDVLARTHTAQDRPPAEGDDRFSESILTYVAHTSALTYPPGPRTQLREDLLPRTLGQLQPDELSPKVDSGVDRHHLMAALSAYAAQRPPAPPGEGSLEPQYLLRAPSRMPRPLLAPAAPQKWPSPLGDSEDPSSTGDGARIHTLLKDLQRQPAEVRGLSGLELDGMAELMAGLMQGVDHGVARGSPGRAALGESGEQADGPKATLRGDSFPDDGVQDDDDRLYQEVHRLSATLGGLLQDHGSRLLPGALPFARPLDMERKKSEHPESSLSSEEETAGVENVKSQTYSKDLLGQQPHSEPGAAAFGELQNQMPGPSKEEQSLPAGAQEALSDGLQLEVQPSEEEARGYIVTDREVLGPAVTFKVSANVQNVTTEDVEKATVDNKDKLEETSGLKILQTGVGSKSKLKFLPPQAEQEDSTKFIALTLVSLACILGVLLASGLIYCLRHSSQHRLKEKLSGLGGDPGADATAAYQELCRQRMATRPPDRPEGPHTSRISSVSSQFSDGPIPSPSARSSASSWSEEPVQSNMDISTGHMILSYMEDHLKNKNRLEKEWEALCAYQAEPNSSFVAQREENVPKNRSLAVLTYDHSRVLLKAENSHSHSDYINASPIMDHDPRNPAYIATQGPLPATVADFWQMVWESGCVVIVMLTPLAENGVRQCYHYWPDEGSNLYHIYEVNLVSEHIWCEDFLVRSFYLKNLQTNETRTVTQFHFLSWYDRGVPSSSRSLLDFRRKVNKCYRGRSCPIIVHCSDGAGRSGTYVLIDMVLNKMAKGAKEIDIAATLEHLRDQRPGMVQTKEQFEFALTAVAEEVNAILKALPQ

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José J Ramírez-Franco et al.
Frontiers in cellular neuroscience, 10, 202-202 (2016-09-16)
Hippocampal interneurons comprise a diverse family of inhibitory neurons that are critical for detailed information processing. Along with gamma-aminobutyric acid (GABA), interneurons secrete a myriad of neuroactive substances via secretory vesicles but the molecular composition and regulatory mechanisms remain largely

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service