Skip to Content
Merck
All Photos(1)

Key Documents

S9070

Sigma-Aldrich

SB 220025 trihydrochloride

≥98% (HPLC), solid

Synonym(s):

5-(2-Aminopyrimidin-4-yl)-4-(4-fluorophenyl)-1-(4-piperidinyl)imidazole trihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$159.00
2 MG
$673.00

$159.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
500 μG
$159.00
2 MG
$673.00

About This Item

Empirical Formula (Hill Notation):
C18H19FN6 · 3HCl
CAS Number:
Molecular Weight:
447.76
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$159.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

color

white

solubility

H2O: 22 mg/mL

originator

GlaxoSmithKline

SMILES string

Cl[H].Cl[H].Cl[H].Nc1nccc(n1)-c2c(ncn2C3CCNCC3)-c4ccc(F)cc4

InChI

1S/C18H19FN6.3ClH/c19-13-3-1-12(2-4-13)16-17(15-7-10-22-18(20)24-15)25(11-23-16)14-5-8-21-9-6-14;;;/h1-4,7,10-11,14,21H,5-6,8-9H2,(H2,20,22,24);3*1H

InChI key

IPFBHUGKKONHFR-UHFFFAOYSA-N

Application

SB 220025 trihydrochloride may be used in p38-mediated cell signaling studies.

Biochem/physiol Actions

SB 220025 is a potent, specific inhibitor of human p38 MAP kinase (IC50 = 60 nM).
SB 220025 is a potent, specific inhibitor of human p38 mitogen-activated protein (MAP) kinase, with an IC50 value of 60 nM and 50- to 1000-fold selectivity. SB 220025 reduces inflammatory cytokine production and inhibits angiogenesis. SB 220025 reduced the lipopolysaccharide-induced production of TNF at an ED50 value of 7.5 mg/kg. At 30 mg/kg, SB 220025 reduced the expression of TNF and inhibited angiogenesis by ~40%. In a further study, the effects of p38/CSBP MAP kinase inhibition in angiogenesis-dependent chronic inflammatory disease was tested in murine collagen-induced arthritis and SB 220025 was able to prevent the progression of established arthritis.

Features and Benefits

This compound is featured on the MAPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service