Skip to Content
Merck
All Photos(1)

Key Documents

559396

Sigma-Aldrich

SB 220025

A cell-permeable, potent, reversible, ATP-competitive, and specific inhibitor of human p38 MAP Kinase (IC₅₀ = 60 nM).

Synonym(s):

SB 220025, 5-(2-Amino-4-pyrimidinyl)-4-(4-fluorophenyl)-1-(4-piperidinlyl)imidazole, p38 MAP Kinase Inhibitor XIV

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$423.00

$423.00


Check Cart for Availability


Select a Size

Change View
500 μG
$423.00

About This Item

Empirical Formula (Hill Notation):
C18H19FN6
CAS Number:
Molecular Weight:
338.38
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

$423.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

solid

potency

60 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white to pale yellow

solubility

DMSO: 22 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Fc1ccc(cc1)c2nc[n](c2c4nc(ncc4)N)C3CCNCC3

InChI

1S/C18H19FN6/c19-13-3-1-12(2-4-13)16-17(15-7-10-22-18(20)24-15)25(11-23-16)14-5-8-21-9-6-14/h1-4,7,10-11,14,21H,5-6,8-9H2,(H2,20,22,24)

InChI key

VSPFURGQAYMVAN-UHFFFAOYSA-N

General description

A cell-permeable, potent, reversible, ATP-competitive, and specific inhibitor of human p38 MAP Kinase (IC50 = 60 nM). Binds to an extended pocket in the active site of the enzyme. Exhibits over 2000-fold greater selectivity for p38 MAPK over ERK (p42/p44 MAP kinase), 500-fold over PKA, 50-fold over PKC, and >1000-fold over EGFR. Also acts as a potent inhibitor of angiogenesis and as an inhibitor of LPS-induced TNF-α production.

Please note that the molecular weight for this compound is batch-specific due to variable water content.
A potent and specific inhibitor of human p38 mitogen-activated protein (MAP) kinase (IC50 = 60 nM). Binds to an extended pocket in the active site of the enzyme. Exhibits >2500-fold greater selectivity for p38 MAPK over ERK (p42/p44 MAP kinase), 500-fold over PKA, 50-fold over PKC, and >1600-fold over EGFRK. Also acts as a potent inhibitor of angiogenesis and as an inhibitor of LPS-induced TNF-α production.

Biochem/physiol Actions

Primary Target
P38MAPK

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Jackson, J.R., et al. 1998. J. Pharmacol. Exp. Ther.284, 687.
Wang, Z., et al. 1998. Structure6, 1117.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service