Skip to Content
Merck
All Photos(1)

Key Documents

RABQUENCH

Sigma-Aldrich

Quenching Solution for Cell-based ELISA Assay (Item E)

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$35.30

$35.30


Check Cart for Availability


Select a Size

Change View
2 ML
$35.30

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$35.30


Check Cart for Availability

shipped in

wet ice

storage temp.

−20°C

Other Notes

This item is for RAB series CBEL ELISA kits.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service