Skip to Content
Merck
All Photos(2)

Key Documents

RAB0407

Sigma-Aldrich

Human Angiostatin ELISA Kit

serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$918.00

$918.00


Check Cart for Availability


Select a Size

Change View
1 KIT
$918.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$918.00


Check Cart for Availability

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20000 pg/mL
standard curve range: 31.25-2000 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PLG(5340)

General description

The Human Angiostatin ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human Angiostatin in serum, plasma (heparin), cell culture supernatants and urine.

Immunogen

Recombinant Human Angiostatin

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Gavriilaki et al.
Journal of human hypertension, 28(10), 606-609 (2014-03-14)
Essential hypertension (EH) is characterised by increased thrombotic tendency and impaired fibrinolytic activity. However, exercise-induced changes in coagulation and fibrinolysis have not yet been clarified. We aimed at determining thrombotic and fibrinolytic activity during exercise in patients with EH pre
Simone Azevedo Zanette et al.
Molecular pain, 10, 46-46 (2014-07-10)
Fibromyalgia (FM) is conceptualized as a central sensitization (CS) condition, that presents high serum brain-derived neurotrophic factor (BDNF) and neuroglia activation. Although the S100B protein regulates neuroglia functions, it has been traditionally used as a proxy of central nervous system
Dawn M Waterworth et al.
Journal of the American Heart Association, 3(4) (2014-08-29)
Genetics can be used to predict drug effects and generate hypotheses around alternative indications. To support Losmapimod, a p38 mitogen-activated protein kinase inhibitor in development for acute coronary syndrome, we characterized gene variation in MAPK11/14 genes by exome sequencing and
Matthew T Balmer et al.
Cancer biology & therapy, 15(8), 1000-1012 (2014-05-08)
The chemotherapeutic agents doxorubicin (dox) or 5-fluorouracil (5FU) are used to treat cancer cells as they cause irreparable DNA damage, inducing these aberrant cells to undergo cell death. The mediator of this process is presumed to be in part the
Ahmed M Abu El-Asrar et al.
Molecular vision, 20, 1209-1224 (2014-09-26)
The calcium-binding protein S100A4 is implicated in cancer cell invasion and metastasis, the stimulation of angiogenesis, the progression of fibrosis, and inflammatory disorders. We investigated the expression of S100A4 and correlated it with clinical disease activity as well as with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service