Skip to Content
Merck
All Photos(1)

Key Documents

GF171

Sigma-Aldrich

Endostatin Protein, Human Recombinant

The Endostatin protein is a naturally occurring 20 kDa polypeptide derived from the C-terminal portion of type XVIII collagen.

Synonym(s):

Collagen Type XVIII, Collagen alpha-1 (XVIII)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$641.00

$641.00


Estimated to ship on25 May 2025



Select a Size

Change View
100 μG
$641.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

$641.00


Estimated to ship on25 May 2025


recombinant

expressed in E. coli

Quality Level

Assay

>98% (SDS-PAGE and HPLC)

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

UniProt accession no.

shipped in

dry ice

General description

Endostatin is a naturally occurring 20 kDa polypeptide derived from the C-terminal portion of type XVIII collagen. It functions as an anti-angiogenic cytokine that is expressed in various organs with the highest levels in liver, lung and kidney. Endostatin inhibits angiogenesis by blocking the pro-angiogenic activities of VEGF and FGF-basic. Recombinant human Endostatin is a 20.2 kDa protein consisting 184 amino acid residues.
Product Source: E. coli.

Specificity

Cross Reactivty
None

Application

Research Category
Stem Cell Research
Research Sub Category
Growth Factors & Receptors

Quality

Bioactivity assay: Measured by its ability to inhibit HUVEC migration.

Physical form

Product is filtered through a 0.2 micron filter before lyophilization.

Storage and Stability

Store at -20°C for up to 6 months from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service