Skip to Content
Merck
All Photos(1)

Key Documents

RAB0386

Sigma-Aldrich

Brain Natriuretic Peptide EIA Kit

for serum, plasma, culture supernatant and cell lysates

Synonym(s):

Gamma-brain natriuretic peptide, Natriuretic peptides B , BNP

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$865.00

$865.00


Check Cart for Availability


Select a Size

Change View
1 KIT
$865.00

About This Item

UNSPSC Code:
41123300
NACRES:
NA.32

$865.00


Check Cart for Availability

species reactivity

human, mouse, rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

input

sample type serum
sample type culture supernatant(s)
sample type plasma
sample type cell lysate

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 1.66 pg/mL
standard curve range: 0.1–1000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NPPB(4879)

General description

The BNP Enzyme Immunoassay (EIA) Kit is an in vitro quantitative assay for detecting BNP peptide based on the principle of Competitive Enzyme Immunoassay.

Immunogen

Brain Natriuretic Peptide, synthetic peptide

Application

For research use only. Not for use in diagnostic procedures.
Brain natriuretic peptide EIA Kit has been used to measure brain natriuretic peptide (BNP) secreted in human neonatal ventricular cardiomyocyte culture.[1][2]

Biochem/physiol Actions

Brain natriuretic peptide (BNP) expressed in mammalian brain and atrial natriuretic peptide (ANP) facilitates various physiological functions including regulation of blood pressure and cardiac remodeling.[3] BNP acts as a potential biomarker of high left ventricular end-diastolic pressure in patients with symptomatic left ventricular (LV) dysfunction.[4] Elevated expression of BNP is observed in acute myocardial infarction (AMI) patients. BNP is also implicated in several biological functions such as diuresis, natriureis, hypotensive action and inhibition of aldosterone secretion.[3]

Kit Components Also Available Separately

Product No.
Description
SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plasma brain natriuretic peptide as a biochemical marker of high left ventricular end-diastolic pressure in patients with symptomatic left ventricular dysfunction.
Maeda K
American Heart Journal, 135(5 pt 1), 825-832 (1998)
Azar Hosseini et al.
Frontiers in pharmacology, 13, 909079-909079 (2022-06-28)
Background: Doxorubicin as an anti-cancer drug causes cardiotoxicity, limiting its tolerability and use. The mechanism of toxicity is due to free radical production and cardiomyocytes injury. This research evaluated Rheum turkestanicum (R.turkestanicum) extract against doxorubicin cardiotoxicity due to its considerable
A new natriuretic peptide in porcine brain.
Sudoh T
Nature, 332(6159), 78-81 (1988)
Distinct T cell signatures define subsets of patients with multiple sclerosis.
Johnson MC
Neurology? neuroimmunology & neuroinflammation, 3(5), e278-e278 (2016)
Jiakan Weng et al.
Journal of cellular and molecular medicine, 23(2), 1050-1058 (2018-11-20)
Obesity is associated with increased cardiovascular morbidity and mortality, but the direct signals to initiate or exaggerate cardiomyopathy remain largely unknown. Present study aims to explore the pathophysiological role of autotaxin/lysophosphatidic acid (LPA) in the process of cardiomyopathy during obesity.

Questions

  1. What is the stability of the anti-BNP antibody concentrate (item N reconstituted with 55 ul of 1X Assay Diluent) from RAB0386 when stored at 4C?

    1 answer
    1. Storing and reusing the reconstituted antibody item N is not recommended as it is not stable when stored. The kit includes 2 vials so that a partial plate may be run.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service