Skip to Content
Merck
All Photos(2)

Documents

RAB0268

Sigma-Aldrich

Human IL-18 R β ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type urine
sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 12 pg/mL
standard curve range: 12.29-3000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL18RAP(8807)

General description

The Human IL-18R β ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL-18R β in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human IL-18 R β

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABWASH420X Wash Buffer (Item B)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADDELISA 5X Assay/Sample Diluent Buffer D (Item K)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zufeng Ding et al.
Biochemical and biophysical research communications, 451(4), 637-643 (2014-08-19)
Lectin-like oxidized low-density lipoprotein scavenger receptor-1 (LOX-1) is known to be involved in many pathophysiological events, such as inflammation. To clarify the role of LOX-1 in mtDNA damage and NLRP3 inflammasome activation, we studied wild-type (WT) and LOX-1 knockout (KO)
Benjamin Uzan et al.
EMBO molecular medicine, 6(6), 821-834 (2014-04-30)
Development of novel therapies is critical for T-cell acute leukaemia (T-ALL). Here, we investigated the effect of inhibiting the MAPK/MEK/ERK pathway on T-ALL cell growth. Unexpectedly, MEK inhibitors (MEKi) enhanced growth of 70% of human T-ALL cell samples cultured on
Wenmei Zhang et al.
International journal of molecular medicine, 45(2), 461-474 (2020-01-03)
The aim of the present study was to investigate the role of microRNA‑15a‑5p (miR‑15a‑5p) in pulmonary arterial hypertension (PAH) and elucidate the underlying pro‑apoptotic mechanism. Reverse transcription‑quantitative PCR analysis and gene microarray hybridization were used to measure the expression of
Kaori Sadakane et al.
Journal of applied toxicology : JAT, 34(8), 893-902 (2013-08-29)
Phthalate esters in plastics act as adjuvants for immunoglobulin production, which aggravates allergic disease. However, the effects of alkylphenols (used as plasticizers and surfactants) on atopic dermatitis have not been studied in detail. Therefore, the goal of the present study
J Michelle Kahlenberg et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(1), 152-162 (2014-01-23)
Systemic lupus erythematosus (SLE) is a systemic autoimmune syndrome associated with organ damage and an elevated risk of cardiovascular disease resulting from activation of both innate and adaptive immune pathways. Recently, increased activation of the inflammasome machinery in SLE has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service