Merck
  • MicroRNA‑15a‑5p induces pulmonary artery smooth muscle cell apoptosis in a pulmonary arterial hypertension model via the VEGF/p38/MMP‑2 signaling pathway.
MicroRNA‑15a‑5p induces pulmonary artery smooth muscle cell apoptosis in a pulmonary arterial hypertension model via the VEGF/p38/MMP‑2 signaling pathway.
International journal of molecular medicine (2020-01-03)
Wenmei Zhang, Yanna Li, Xin Xi, Guangfa Zhu, Shenghao Wang, Yan Liu, Man Song
ABSTRACT
The aim of the present study was to investigate the role of microRNA‑15a‑5p (miR‑15a‑5p) in pulmonary arterial hypertension (PAH) and elucidate the underlying pro‑apoptotic mechanism. Reverse transcription‑quantitative PCR analysis and gene microarray hybridization were used to measure the expression of miR‑15a‑5p in the lung tissues of rats with monocrotaline (MCT)‑induced PAH. Flow cytometry and caspase‑3/9 activity assays were adopted to measure the apoptosis of pulmonary artery smooth muscle cells (PASMCs). The expression of apoptosis‑related proteins was analyzed using western blotting. The results demonstrated that the expression of miR‑15a‑5p was significantly increased in the lung tissues of rats with MCT‑induced PAH. In addition, the overexpression of miR‑15a‑5p reduced PASMC proliferation, induced apoptosis, promoted the activity of caspase‑3/9, induced the protein expression of B‑cell lymphoma 2‑associated X protein (Bax), decreased the expression of B‑cell lymphoma 2 (Bcl‑2), increased inflammation, as indicated by the expression of tumor necrosis factor‑α (TNF)‑α and interleukin (IL)‑1β, IL‑6 and IL‑18, suppressed the protein expression of vascular endothelial growth factor (VEGF), and promoted the protein expression levels of phosphorylated (p)‑p38 mitogen‑activated protein kinase (p38) and matrix metalloproteinase (MMP)‑2 in the PASMCs of rats with MCT‑induced PAH. By contrast, the downregulation of miR‑15a‑5p increased cell proliferation, decreased apoptosis, reduced the activity of caspase‑3/9 and the protein expression of Bax, increased the expression of Bcl‑2, inhibited inflammation (as suggested by the expression of TNF‑α, IL‑1β, IL‑6 and IL‑18), induced the protein expression of VEGF, and suppressed the protein expression of p‑p38 and MMP‑2 in the PASMCs of rats with MCT‑induced PAH. The inhibition of VEGF attenuated the effects of the overexpression of miR‑15a‑5p on the inhibition of cell proliferation, apoptotic rate, caspase‑3/9 activity and protein expression of Bax, and it attenuated the increased inflammation, as indicated by the protein expression of p38 and MMP‑2 in the PASMCs. In conclusion, the data of the present study demonstrated that miR‑15a‑5p induced the apoptosis of PASMCs in an animal model of PAH via the VEGF/p38/MMP‑2 signaling pathway. However, further research is required to fully elucidate the role of miR‑15a‑5p in the development of PAH.
Sigma-Aldrich
Human IL-18 R β ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Mouse Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-1 β ELISA Kit, for serum, plasma and cell culture supernatant