Skip to Content
Merck
All Photos(2)

Key Documents

RAB0196

Sigma-Aldrich

Human FLRG ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,320.00

$1,320.00


Check Cart for Availability


Select a Size

Change View
1 KIT
$1,320.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$1,320.00


Check Cart for Availability

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type urine
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 250 pg/mL
standard curve range: 204.8-50000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FSTL3(10272)

General description

The Human FLRG ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human Follistatin-related Gene product

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiamin Xie et al.
Frontiers in cell and developmental biology, 8, 573781-573781 (2020-11-17)
Growth differentiation factor 8 (GDF8) and its antagonist follistatin-like 3 (FSTL3) are expressed in the placenta during early pregnancy. These two factors may have a role to play in the regulation of normal placentation. However, whether GDF8 can regulate the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service