Skip to Content
Merck
All Photos(1)

Key Documents

RABELADC

Sigma-Aldrich

ELISA 1X Assay/Sample Diluent Buffer C (Item L)

Sign Into View Organizational & Contract Pricing

Select a Size

30 ML
$130.00

$130.00


Check Cart for Availability


Select a Size

Change View
30 ML
$130.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$130.00


Check Cart for Availability

shipped in

wet ice

storage temp.

−20°C

Other Notes

Intended for use with ELISA kits.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service