RAB0012
Phospho-Akt (pSer473) / pan-Akt ELISA Kit
for detection of human, mouse or rat phospho-akt (pSer473) and pan-akt in cell and tissue lysates
Synonym(s):
Akt ELISA Kit
About This Item
Recommended Products
species reactivity
human, mouse, rat
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type: human cell lysate
sample type tissue lysate
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... AKT1(207)
Related Categories
General description
Immunogen
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Kit Components Also Available Separately
- RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service