Skip to Content
Merck
All Photos(1)

Key Documents

R7532

Sigma-Aldrich

RRD-251 hydrochloride

≥98% (HPLC)

Synonym(s):

(2,4-Dichlorophenyl)carbamimidothioic acid methyl ester hydrochloride, 2-(2,4-Dichlorobenzyl)-2-thiopseudourea hydrochloride, Rb/Raf-1 disruptor 251 hydrochloride, S-(2,4-Dichlorobenzyl)-isothiouronium hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$43.70
25 MG
$176.00

$43.70


Estimated to ship on28 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$43.70
25 MG
$176.00

About This Item

Empirical Formula (Hill Notation):
C8H8Cl2N2S· HCl
CAS Number:
Molecular Weight:
271.59
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$43.70


Estimated to ship on28 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: >10 mg/mL
H2O: >2 mg/mL

storage temp.

room temp

SMILES string

Cl.NC(=N)SCc1ccc(Cl)cc1Cl

InChI

1S/C8H8Cl2N2S.ClH/c9-6-2-1-5(7(10)3-6)4-13-8(11)12;/h1-3H,4H2,(H3,11,12);1H

InChI key

COMNQRICZGJVLE-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

RRD-251 hydrochloride is a reversible, potent, and selective disruptor of Rb/Raf-1 interaction.
RRD-251 hydrochloride is a reversible, potent, and selective disruptor of Rb/Raf-1 interaction. The retinoblastoma tumor suppressor protein (Rb) controls the G1-S boundary by repressing the transcriptional activity of the E2F family of transcription factors. Raf-1 kinase binds and phosphorylates Rb early in the G1 phase. RRD-251 significantly inhibits angiogenesis and tumor growth in vivo in an Rb-dependent manner. RRD-251 does not inhibit the binding of B-Raf to Rb and Raf-1 to Mek1/2. Also, RRD-251 does not affect the kinase activities associated with cyclin D, cyclin E, or Raf-1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandeep Singh et al.
Molecular cancer therapeutics, 9(12), 3330-3341 (2010-12-09)
Metastatic melanoma is an aggressive cancer with very low response rate against conventional chemotherapeutic agents such as dacarbazine (DTIC). Inhibitor of Rb-Raf-1 interaction RRD-251 was tested against the melanoma cell lines SK-MEL-28, SK-MEL-5, and SK-MEL-2. RRD-251 was found to be
Debora Stelitano et al.
Oncotarget, 8(40), 67422-67438 (2017-10-06)
GTSE1 over-expression has been reported as a potential marker for metastasis in various types of malignancies, including breast cancer. Despite this, the transcriptional regulation of this protein and the causes of its misregulation in tumors remain largely unknown. The aims

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service