Skip to Content
Merck
All Photos(1)

Key Documents

8.14145

Sigma-Aldrich

Ethylene glycol bis-mercaptoacetate

for synthesis

Synonym(s):

Ethylene glycol bis-mercaptoacetate, Ethylene glycol dithioacetate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$116.00

$116.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$116.00

About This Item

Linear Formula:
(HSCH2COOCH2)2
CAS Number:
Molecular Weight:
210.27
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-653-4
NACRES:
NA.22

$116.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

form

liquid

potency

330 mg/kg LD50, oral (Rat)

bp

137-139 °C/2 hPa

density

1.32 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

SCC(=O)OCCOC(=O)CS

InChI

1S/C6H10O4S2/c7-5(3-11)9-1-2-10-6(8)4-12/h11-12H,1-4H2

InChI key

PSYGHMBJXWRQFD-UHFFFAOYSA-N

Analysis Note

Assay (iodometric): ≥ 95.0 %
Density (d 20 °C/ 4 °C): 1.310 - 1.320
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service