Skip to Content
Merck
All Photos(1)

Key Documents

R6391

Sigma-Aldrich

Unwire full racks, polyketone

Holds 40 x 25-mm tubes, white

Synonym(s):

moulded rack, test tube rack

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$86.20

$86.20


Estimated to ship on19 May 2025



Select a Size

Change View
1 EA
$86.20

About This Item

UNSPSC Code:
41122804
NACRES:
NB.35

$86.20


Estimated to ship on19 May 2025


material

polyketone

feature

Holds 40 x 25-mm tubes

manufacturer/tradename

Nalgene® 5970-0025

L × W × H

300 mm × 121 mm × 92 mm

color

white

Looking for similar products? Visit Product Comparison Guide

General description

Molded of durable polyketone. Unwire test tube racks have no coating that can chip or peel and permit corrosion. Excellent replacement for wire racks; withstands most lab chemicals. Excellent thermal properties and won′t get brittle in freezers. Grid design allows tubes to be tipped from side to side for clearly marking position. These racks will not float in water baths.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Unwire is a trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service