Skip to Content
Merck
All Photos(1)

Key Documents

Z740270

Aluminum heating/cooling block

Aluminum heating/cooling block

Holds 96 x 0.2 mL tubes in center plus 10 on size (4 for 1.5ml tubes, 6 for 0.5ml tubes)

Synonym(s):

dry block, tube cooling block, tube heating block

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$390.00

$390.00


Check Cart for Availability


Select a Size

Change View
1 EA
$390.00

About This Item

UNSPSC Code:
41102404
NACRES:
NB.35

$390.00


Check Cart for Availability

material

aluminum

feature

Holds 96 x 0.2 mL tubes in center plus 10 on size (4 for 1.5ml tubes, 6 for 0.5ml tubes)

packaging

pkg of 1 ea

manufacturer/tradename

Diversified Biotech CHAM-1000

technique(s)

PCR: suitable

size

156x76x25 mm

Looking for similar products? Visit Product Comparison Guide

General description

Each Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures.

Application

Aluminum heating/cooling block has been used to cool PCR tubes during dilution of DNA fragments and the preparation of the enzyme master mix for in vitro cyclic GMP-AMP synthase (cGAS)reaction.[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Andreas Holleufer et al.
Bio-protocol, 8(20), e3055-e3055 (2018-10-20)
Cyclic GMP-AMP synthase (cGAS) is a pattern recognition receptor (PRR) that senses double stranded DNA (dsDNA) in the cytosol and this leads to the activation of stimulator of interferon genes (STING) via the secondary messenger 2'3'-cyclic GMP-AMP (2'3'-cGAMP). STING then

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service