Skip to Content
Merck
All Photos(1)

Key Documents

R3251

Sigma-Aldrich

D-Ribulose-5-phosphate 3-Epimerase from baker′s yeast (S. cerevisiae)

lyophilized powder, 50-100 units/mg protein (modified Warburg-Christian)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

biological source

bakers yeast

form

lyophilized powder

specific activity

50-100 units/mg protein (modified Warburg-Christian)

foreign activity

phosphoriboisomerase, alcohol dehydrogenase, transketolase, and transaldolase <0.1%

storage temp.

−20°C

Application

D-Ribulose-5-phosphate 3-Epimerase is an enzyme that converts the reversible conversion of D-ribulose 5-phosphate into D-xylulose 5-phosphate, which is important for the cellular response against oxidative stress [1]. D-Ribulose-5-phosphate 3-Epimerase is involved in the pentose phosphate pathway, pentose and glucuronate interconversions and carbon fixation. Product R3251 is from baker′s yeast and is provided as a lyophilized powder. It is useful in enzyme systems requiring low sulfate.

Biochem/physiol Actions

RPE is a metalloenzyme and has been shown to use the divalent Zn2+ ion predominantly for catalysis. Human D-ribulose-5-phosphate 3-epimerase (hRPE) has been shown to use Fe2+ for catalysis [1].

Unit Definition

One unit will convert 1 μmole of D-ribulose 5-phosphate to D-xylulose 5-phosphate per min at pH 7.7 at 25°C when coupled with transketolase, α-glycerophosphate dehydrogenase, and triosephosphate isomerase.

Physical form

Lyophilized and essentially sulfate-free; contains approx. 35% citrate buffer salts

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Teige et al.
FEBS letters, 377(3), 349-352 (1995-12-27)
A cDNA clone encoding the chloroplast enzyme pentose-5-phosphate 3-epimerase (EC 5.1.3.1) in potato (Solanum tuberosum) was isolated and sequenced. The deduced sequence of 235 amino acids is similar to protein sequences of bacterial epimerases. Northern blot analysis showed the highest
Matthew B Rogers et al.
BMC evolutionary biology, 7, 89-89 (2007-06-15)
Lateral gene transfer is increasingly invoked to explain phylogenetic results that conflict with our understanding of organismal relationships. In eukaryotes, the most common observation interpreted in this way is the appearance of a bacterial gene (one that is not clearly
J Caruthers et al.
Proteins, 62(2), 338-342 (2005-11-24)
The crystal structure of Pfal009167AAA, a putative ribulose 5-phosphate 3-epimerase (PfalRPE) from Plasmodium falciparum, has been determined to 2 A resolution. RPE represents an exciting potential drug target for developing antimalarials because it is involved in the shikimate and the
Kui K Chan et al.
Biochemistry, 47(36), 9608-9617 (2008-08-15)
Enzymes that share the (beta/alpha) 8-barrel fold catalyze a diverse range of reactions. Many utilize phosphorylated substrates and share a conserved C-terminal (beta/alpha) 2-quarter barrel subdomain that provides a binding motif for the dianionic phosphate group. We recently reported functional
Helena Sävenstrand et al.
Plant & cell physiology, 43(4), 402-410 (2002-04-30)
Suppression subtractive hybridisation was used to isolate genes differentially regulated by low levels (UV-B(BE,300) 0.13 W m(-2)) of ultraviolet-B radiation (UV-B; 290-320 nm) in Pisum sativum. Six genes were regulated, two of which were novel. The mRNA levels for these

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service