Skip to Content
Merck
All Photos(5)

Documents

G0380

Sigma-Aldrich

α-D-Galactose 1-phosphate dipotassium salt pentahydrate

Type II, ≥98% (HPLC)

Synonym(s):

α-D-Galactopyranosyl 1-phosphate dipotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11K2O9P · 5H2O
CAS Number:
Molecular Weight:
426.39
Beilstein:
3922740
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

bovine milk

type

Type II

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

impurities

≤0.5 mol % glucose 1-phosphate.

color

white

solubility

water: 50 mg/mL, clear, colorless

cation traces

K: 20.4-26.1% (anhydrous)

storage temp.

−20°C

SMILES string

[K+].[K+].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C6H13O9P.2K.5H2O/c7-1-2-3(8)4(9)5(10)6(14-2)15-16(11,12)13;;;;;;;/h2-10H,1H2,(H2,11,12,13);;;5*1H2/q;2*+1;;;;;/p-2/t2-,3+,4+,5-,6-;;;;;;;/m1......./s1

InChI key

ZYBRMLNCQMITLC-JFYWUTKTSA-L

Looking for similar products? Visit Product Comparison Guide

Application

α-D-Galactose 1-phosphate may be used in Leloir pathway of galactose metabolism research to study the process of UDP-Galactose (UDP-Gal) formation via UDP-glucose:α-D-galactose-1-phosphate uridylyltransferase(s) (EC 2.7.7.12). It may be used to identify and characterize and study the functions of galactose-1-phosphate uridylyltransferase(s).

Biochem/physiol Actions

Metabolite intermediate in the interconversion of glucose and galactose. Also involved in galactose metabolism and nucleotide sugar metabolism

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service